Assay Design



Gene Transcripts

Gene Symbol
TEX9
Entrez Gene ID
374618
Gene Name
testis expressed 9
Gene Aliases
-
Chromosome Location
Chr.15: 56243971 - 56445997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286449.1 NP_001273378.1 3 - 4 438 67
NM_198524.2 NP_940926.1 3 - 4 215 67
XM_005254359.4 XP_005254416.1 3 - 4 385 67
XM_005254361.3 XP_005254418.1 2 - 3 377 67
XM_011521530.2 XP_011519832.1 3 - 4 385 67
XM_017022160.1 XP_016877649.1 3 - 4 385 67
XM_017022161.1 XP_016877650.1 3 - 4 264 67
XM_017022162.1 XP_016877651.1 3 - 4 264 67
XM_017022163.1 XP_016877652.1 2 - 3 477 67
XM_017022164.1 XP_016877653.1 3 - 4 436 67
XM_017022166.1 XP_016877655.1 2 - 3 288 67
XM_017022167.1 XP_016877656.1 3 - 4 436 67
GenBank mRNA AK294960.1 - 3 - 4 424 67
AL832050.1 - 2 - 3 294 67
BC028119.1 - 3 - 4 193 67
HY164049.1 - 3 - 4 215 67
KU178902.1 - 3 - 4 188 67
KU178903.1 - 3 - 4 411 67

View Full Product Details