Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286449.1 | NP_001273378.1 | 6 - 7 | 644 | 134 |
NM_198524.2 | NP_940926.1 | 6 - 7 | 421 | 134 | |
XM_005254359.4 | XP_005254416.1 | 6 - 7 | 591 | 134 | |
XM_005254361.3 | XP_005254418.1 | 5 - 6 | 583 | 134 | |
XM_011521530.2 | XP_011519832.1 | 6 - 7 | 591 | 134 | |
XM_017022160.1 | XP_016877649.1 | 6 - 7 | 591 | 134 | |
XM_017022162.1 | XP_016877651.1 | 6 - 7 | 470 | 134 | |
XM_017022163.1 | XP_016877652.1 | 5 - 6 | 683 | 134 | |
XM_017022164.1 | XP_016877653.1 | 6 - 7 | 642 | 134 | |
XM_017022167.1 | XP_016877656.1 | 6 - 7 | 642 | 134 | |
GenBank mRNA | AK294960.1 | - | 6 - 7 | 630 | 134 |
AL832050.1 | - | 5 - 6 | 500 | 134 | |
BC028119.1 | - | 6 - 7 | 399 | 134 | |
HY164049.1 | - | 6 - 7 | 421 | 134 | |
KU178902.1 | - | 6 - 7 | 394 | 134 | |
KU178903.1 | - | 6 - 7 | 617 | 134 |