Assay Design



Gene Transcripts

Gene Symbol
TEX9
Entrez Gene ID
374618
Gene Name
testis expressed 9
Gene Aliases
-
Chromosome Location
Chr.15: 56243971 - 56445997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286449.1 NP_001273378.1 10 - 11 1217 137
NM_198524.2 NP_940926.1 10 - 11 994 137
XM_005254359.4 XP_005254416.1 10 - 11 1164 137
XM_005254361.3 XP_005254418.1 9 - 10 1156 137
XM_011521530.2 XP_011519832.1 10 - 11 1164 137
XM_017022160.1 XP_016877649.1 10 - 11 1164 137
XM_017022161.1 XP_016877650.1 8 - 9 911 137
XM_017022163.1 XP_016877652.1 9 - 10 1256 137
XM_017022164.1 XP_016877653.1 10 - 11 1215 137
XM_017022166.1 XP_016877655.1 7 - 8 935 137
GenBank mRNA AK294960.1 - 10 - 11 1203 137
BC028119.1 - 10 - 11 972 137
KU178902.1 - 10 - 11 967 137
KU178903.1 - 10 - 11 1190 137

View Full Product Details