Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316350.1 | NP_001303279.1 | 10 - 11 | 1617 | 126 |
NM_014236.3 | NP_055051.1 | 11 - 12 | 1800 | 126 | |
XM_005273313.4 | XP_005273370.1 | 11 - 12 | 1760 | 126 | |
XM_011544303.2 | XP_011542605.1 | 12 - 13 | 1849 | 126 | |
XM_011544304.2 | XP_011542606.1 | 11 - 12 | 1668 | 126 | |
GenBank mRNA | AB209882.1 | - | 11 - 12 | 1705 | 126 |
AF035285.1 | - | 5 - 6 | 703 | 126 | |
AF043937.1 | - | 11 - 12 | 1676 | 126 | |
AJ002190.1 | - | 11 - 12 | 1751 | 126 | |
AK222773.1 | - | 11 - 12 | 1756 | 126 | |
AK223304.1 | - | 11 - 12 | 1759 | 126 | |
AK297982.1 | - | 10 - 11 | 1578 | 126 | |
AK311386.1 | - | 10 - 11 | 1450 | 126 | |
AK314181.1 | - | 11 - 12 | 1763 | 126 | |
BC000450.2 | - | 11 - 12 | 1647 | 126 |