Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316350.1 | NP_001303279.1 | 12 - 13 | 1865 | 101 |
NM_014236.3 | NP_055051.1 | 13 - 14 | 2048 | 101 | |
XM_005273313.4 | XP_005273370.1 | 13 - 14 | 2008 | 101 | |
XM_011544303.2 | XP_011542605.1 | 14 - 15 | 2097 | 101 | |
XM_011544304.2 | XP_011542606.1 | 13 - 14 | 1916 | 101 | |
GenBank mRNA | AB209882.1 | - | 13 - 14 | 1953 | 101 |
AF035285.1 | - | 7 - 8 | 951 | 101 | |
AF043937.1 | - | 13 - 14 | 1924 | 101 | |
AJ002190.1 | - | 13 - 14 | 1999 | 101 | |
AK222773.1 | - | 13 - 14 | 2004 | 101 | |
AK297982.1 | - | 12 - 13 | 1826 | 101 | |
AK314181.1 | - | 13 - 14 | 2011 | 101 | |
BC000450.2 | - | 13 - 14 | 1895 | 101 |