Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316350.1 | NP_001303279.1 | 2 - 3 | 464 | 147 |
NM_014236.3 | NP_055051.1 | 3 - 4 | 647 | 147 | |
XM_005273313.4 | XP_005273370.1 | 3 - 4 | 607 | 147 | |
XM_011544303.2 | XP_011542605.1 | 4 - 5 | 696 | 147 | |
XM_011544304.2 | XP_011542606.1 | 3 - 4 | 515 | 147 | |
GenBank mRNA | AB209882.1 | - | 3 - 4 | 552 | 147 |
AF043937.1 | - | 3 - 4 | 523 | 147 | |
AJ002190.1 | - | 3 - 4 | 598 | 147 | |
AK222773.1 | - | 3 - 4 | 603 | 147 | |
AK223304.1 | - | 3 - 4 | 606 | 147 | |
AK297982.1 | - | 2 - 3 | 425 | 147 | |
AK309994.1 | - | 4 - 5 | 692 | 147 | |
AK311386.1 | - | 2 - 3 | 297 | 147 | |
AK314181.1 | - | 3 - 4 | 610 | 147 | |
BC000450.2 | - | 3 - 4 | 494 | 147 | |
DC366380.1 | - | 2 - 3 | 451 | 147 |