Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316350.1 | NP_001303279.1 | 3 - 4 | 591 | 73 |
NM_014236.3 | NP_055051.1 | 4 - 5 | 774 | 73 | |
XM_005273313.4 | XP_005273370.1 | 4 - 5 | 734 | 73 | |
XM_011544303.2 | XP_011542605.1 | 5 - 6 | 823 | 73 | |
XM_011544304.2 | XP_011542606.1 | 4 - 5 | 642 | 73 | |
GenBank mRNA | AB209882.1 | - | 4 - 5 | 679 | 73 |
AF043937.1 | - | 4 - 5 | 650 | 73 | |
AJ002190.1 | - | 4 - 5 | 725 | 73 | |
AK222773.1 | - | 4 - 5 | 730 | 73 | |
AK223304.1 | - | 4 - 5 | 733 | 73 | |
AK297982.1 | - | 3 - 4 | 552 | 73 | |
AK309994.1 | - | 5 - 6 | 819 | 73 | |
AK311386.1 | - | 3 - 4 | 424 | 73 | |
AK314181.1 | - | 4 - 5 | 737 | 73 | |
BC000450.2 | - | 4 - 5 | 621 | 73 |