Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032408.3 | NP_115784.1 | 14 - 15 | 3930 | 90 |
XM_017012773.1 | XP_016868262.1 | 14 - 15 | 3943 | 90 | |
GenBank mRNA | AF072810.1 | - | 14 - 15 | 3930 | 90 |
AF084479.1 | - | 14 - 15 | 3862 | 90 | |
BC136520.1 | - | 14 - 15 | 3763 | 90 |