Assay Design



Gene Transcripts

Gene Symbol
BAZ1B
Entrez Gene ID
9031
Gene Name
bromodomain adjacent to zinc finger domain 1B
Gene Aliases
WBSCR10, WBSCR9, WSTF
Chromosome Location
Chr.7: 73440398 - 73522298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032408.3 NP_115784.1 15 - 16 4083 59
XM_017012773.1 XP_016868262.1 15 - 16 4096 59
GenBank mRNA AB032253.1 - 15 - 16 4076 59
AF072810.1 - 15 - 16 4083 59
AF084479.1 - 15 - 16 4015 59
BC136520.1 - 15 - 16 3916 59

View Full Product Details