Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032408.3 | NP_115784.1 | 4 - 5 | 928 | 70 |
XM_017012773.1 | XP_016868262.1 | 4 - 5 | 941 | 70 | |
GenBank mRNA | AB032253.1 | - | 4 - 5 | 921 | 70 |
AF072810.1 | - | 4 - 5 | 928 | 70 | |
AF084479.1 | - | 4 - 5 | 860 | 70 | |
BC065029.1 | - | 5 - 6 | 630 | 70 | |
BC136520.1 | - | 4 - 5 | 761 | 70 |