Assay Design



Gene Transcripts

Gene Symbol
DGKD
Entrez Gene ID
8527
Gene Name
diacylglycerol kinase delta
Gene Aliases
DGKdelta, dgkd-2
Chromosome Location
Chr.2: 233354507 - 233472098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003648.2 NP_003639.2 11 - 12 1363 73
NM_152879.2 NP_690618.2 12 - 13 1428 73
XM_005246107.3 XP_005246164.1 10 - 11 1095 73
XM_005246108.1 XP_005246165.1 10 - 11 1249 73
XM_011512028.2 XP_011510330.1 12 - 13 2887 73
XM_011512029.2 XP_011510331.1 12 - 13 2888 73
XM_011512030.2 XP_011510332.1 12 - 13 1484 73
XM_011512031.2 XP_011510333.1 12 - 13 1358 73
XM_011512032.2 XP_011510334.1 12 - 13 1342 73
XM_011512033.2 XP_011510335.1 12 - 13 2889 73
XM_011512035.1 XP_011510337.1 10 - 11 1229 73
XM_011512037.2 XP_011510339.1 10 - 11 1333 73
XM_011512039.2 XP_011510341.1 12 - 13 2889 73
XM_011512040.2 XP_011510342.1 10 - 11 1265 73
XM_017005138.1 XP_016860627.1 12 - 13 1766 73
XM_017005139.1 XP_016860628.1 10 - 11 1258 73
XM_017005140.1 XP_016860629.1 12 - 13 1360 73
GenBank mRNA AB078966.1 - 12 - 13 1428 73
AK308846.1 - 10 - 11 1249 73
AK311044.1 - 10 - 11 1182 73
D63479.3 - 12 - 13 1369 73
D73409.1 - 11 - 12 1364 73

View Full Product Details