Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003648.2 | NP_003639.2 | 1 - 2 | 214 | 74 |
NM_152879.2 | NP_690618.2 | 2 - 3 | 279 | 74 | |
XM_011512028.2 | XP_011510330.1 | 2 - 3 | 1738 | 74 | |
XM_011512029.2 | XP_011510331.1 | 2 - 3 | 1739 | 74 | |
XM_011512030.2 | XP_011510332.1 | 2 - 3 | 335 | 74 | |
XM_011512031.2 | XP_011510333.1 | 2 - 3 | 209 | 74 | |
XM_011512032.2 | XP_011510334.1 | 2 - 3 | 193 | 74 | |
XM_011512033.2 | XP_011510335.1 | 2 - 3 | 1740 | 74 | |
XM_011512039.2 | XP_011510341.1 | 2 - 3 | 1740 | 74 | |
XM_017005138.1 | XP_016860627.1 | 2 - 3 | 617 | 74 | |
GenBank mRNA | AB078966.1 | - | 2 - 3 | 279 | 74 |
D63479.3 | - | 2 - 3 | 220 | 74 | |
D73409.1 | - | 1 - 2 | 215 | 74 |