Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016828.2 | NP_058437.1 | 7 - 8 | 1396 | 76 |
NM_016829.2 | NP_058438.1 | 7 - 8 | 1349 | 76 | |
XM_017006498.1 | XP_016861987.1 | 6 - 7 | 1193 | 76 | |
GenBank mRNA | AB019530.1 | - | 6 - 7 | 812 | 76 |
AB019531.1 | - | 6 - 7 | 765 | 76 |