Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005957.4 | NP_005948.3 | 5 - 6 | 1012 | 82 |
XM_005263458.3 | XP_005263515.1 | 5 - 6 | 1849 | 82 | |
XM_005263460.4 | XP_005263517.1 | 5 - 6 | 1467 | 82 | |
XM_005263462.4 | XP_005263519.1 | 5 - 6 | 900 | 82 | |
XM_005263463.3 | XP_005263520.1 | 5 - 6 | 1029 | 82 | |
XM_011541495.2 | XP_011539797.1 | 5 - 6 | 1846 | 82 | |
XM_011541496.2 | XP_011539798.1 | 5 - 6 | 1849 | 82 | |
XM_017001328.1 | XP_016856817.1 | 5 - 6 | 1849 | 82 | |
GenBank mRNA | AB209113.1 | - | 4 - 5 | 1050 | 82 |
AJ237672.1 | - | 4 - 5 | 912 | 82 | |
AK312907.1 | - | 5 - 6 | 873 | 82 | |
BC053509.1 | - | 5 - 6 | 834 | 82 | |
CN355286.1 | - | 1 - 2 | 145 | 82 | |
JQ429447.1 | - | 1 - 2 | 288 | 82 |