Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015074.3 | NP_055889.2 | 10 - 11 | 1193 | 72 |
NM_183416.3 | NP_904325.2 | 10 - 11 | 1193 | 72 | |
GenBank mRNA | AB011163.2 | - | 11 - 12 | 1160 | 72 |
AB017133.1 | - | 10 - 11 | 1043 | 72 | |
AB040881.2 | - | 10 - 11 | 1252 | 72 | |
AB088210.1 | - | 10 - 11 | 959 | 72 | |
AB088211.1 | - | 10 - 11 | 959 | 72 | |
AB088212.1 | - | 10 - 11 | 959 | 72 | |
AB088213.1 | - | 10 - 11 | 959 | 72 | |
AF257176.1 | - | 10 - 11 | 1094 | 72 | |
AY043362.1 | - | 9 - 10 | 945 | 72 | |
AY139835.1 | - | 10 - 11 | 1094 | 72 | |
BC115395.2 | - | 9 - 10 | 1029 | 72 |