Assay Design



Gene Transcripts

Gene Symbol
ARL14
Entrez Gene ID
80117
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
ARF7
Chromosome Location
Chr.3: 160677160 - 160678448 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025047.2 NP_079323.1 1 - 1 1154 73
GenBank mRNA AK026248.1 - 1 - 1 1155 73
BC034354.1 - 1 - 1 1137 73

View Full Product Details