Assay Design



Gene Transcripts

Gene Symbol
EXOC8
Entrez Gene ID
149371
Gene Name
exocyst complex component 8
Gene Aliases
EXO84, Exo84p, SEC84
Chromosome Location
Chr.1: 231332731 - 231337872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175876.4 NP_787072.2 1 - 1 4811 125
GenBank mRNA BM998007.1 - 1 - 1 125
CA775845.1 - 1 - 1 125

View Full Product Details