Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004446.2 | NP_004437.2 | 12 - 13 | 1765 | 96 |
GenBank mRNA | AK296755.1 | - | 12 - 13 | 1712 | 96 |
AK307534.1 | - | 10 - 11 | 1339 | 96 | |
AY493416.1 | - | 10 - 11 | 1312 | 96 | |
BC034797.1 | - | 12 - 13 | 1561 | 96 | |
BC058921.1 | - | 12 - 13 | 1602 | 96 | |
BC126275.1 | - | 12 - 13 | 1633 | 96 | |
BC136465.1 | - | 12 - 13 | 1533 | 96 | |
CR933648.1 | - | 12 - 13 | 1604 | 96 | |
JQ080075.1 | - | 12 - 13 | 1495 | 96 | |
X07466.1 | - | 8 - 9 | 995 | 96 | |
X54326.1 | - | 10 - 11 | 1337 | 96 |