Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004446.2 | NP_004437.2 | 25 - 26 | 3825 | 93 |
XM_017000614.1 | XP_016856103.1 | 14 - 15 | 2081 | 93 | |
GenBank mRNA | AK298192.1 | - | 6 - 7 | 649 | 93 |
AY493416.1 | - | 23 - 24 | 3372 | 93 | |
BC126275.1 | - | 25 - 26 | 3693 | 93 | |
BC136465.1 | - | 25 - 26 | 3593 | 93 | |
X54326.1 | - | 23 - 24 | 3397 | 93 |