Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 17 - 18 | 2187 | 155 |
XM_005268629.3 | XP_005268686.1 | 17 - 18 | 2303 | 155 | |
XM_011537877.2 | XP_011536179.1 | 17 - 18 | 2303 | 155 | |
XM_011537881.2 | XP_011536183.1 | 17 - 18 | 2303 | 155 | |
XM_011537882.2 | XP_011536184.1 | 17 - 18 | 2303 | 155 | |
XM_017018786.1 | XP_016874275.1 | 17 - 18 | 2303 | 155 | |
XM_017018789.1 | XP_016874278.1 | 17 - 18 | 2303 | 155 | |
GenBank mRNA | AK127729.1 | - | 8 - 9 | 917 | 155 |
AY792511.1 | - | 17 - 18 | 2066 | 155 | |
BC117180.1 | - | 17 - 18 | 2124 | 155 | |
BM285390.1 | - | 13 - 14 | 1414 | 155 |