Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 4 - 5 | 557 | 133 |
XM_005268629.3 | XP_005268686.1 | 4 - 5 | 673 | 133 | |
XM_011537877.2 | XP_011536179.1 | 4 - 5 | 673 | 133 | |
XM_011537881.2 | XP_011536183.1 | 4 - 5 | 673 | 133 | |
XM_011537882.2 | XP_011536184.1 | 4 - 5 | 673 | 133 | |
XM_017018786.1 | XP_016874275.1 | 4 - 5 | 673 | 133 | |
XM_017018789.1 | XP_016874278.1 | 4 - 5 | 673 | 133 | |
GenBank mRNA | AY792511.1 | - | 4 - 5 | 436 | 133 |
BC117180.1 | - | 4 - 5 | 494 | 133 | |
BM285390.1 | - | 4 - 5 | 326 | 133 |