Assay Design



Gene Transcripts

Gene Symbol
CLIC3
Entrez Gene ID
9022
Gene Name
chloride intracellular channel 3
Gene Aliases
-
Chromosome Location
Chr.9: 136994608 - 136997929 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004669.2 NP_004660.2 1 - 2 63 103
GenBank mRNA BC007012.1 - 1 - 2 59 103
BM043659.1 - 1 - 2 294 103

View Full Product Details