Assay Design



Gene Transcripts

Gene Symbol
OSCP1
Entrez Gene ID
127700
Gene Name
organic solute carrier partner 1
Gene Aliases
C1orf102, NOR1
Chromosome Location
Chr.1: 36417906 - 36450485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145047.4 NP_659484.4 4 - 5 632 94
XM_005270461.1 XP_005270518.1 5 - 6 662 94
XM_005270462.1 XP_005270519.1 5 - 6 662 94
XM_011540680.2 XP_011538982.1 6 - 7 704 94
XM_011540681.2 XP_011538983.1 5 - 6 555 94
XM_017000303.1 XP_016855792.1 4 - 5 568 94
XM_017000304.1 XP_016855793.1 4 - 5 568 94
GenBank mRNA AB079075.1 - 4 - 5 516 94
AF462348.1 - 4 - 5 674 94
AK098541.1 - 5 - 6 628 94
BC018069.1 - 4 - 5 598 94
BF691288.1 - - 421 94

View Full Product Details