Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145047.4 | NP_659484.4 | 7 - 8 | 933 | 76 |
XM_005270461.1 | XP_005270518.1 | 8 - 9 | 963 | 76 | |
XM_011540680.2 | XP_011538982.1 | 9 - 10 | 1005 | 76 | |
XM_011540681.2 | XP_011538983.1 | 8 - 9 | 856 | 76 | |
GenBank mRNA | AB079075.1 | - | 7 - 8 | 817 | 76 |
AF462348.1 | - | 7 - 8 | 975 | 76 | |
AK098541.1 | - | 8 - 9 | 929 | 76 | |
BC018069.1 | - | 7 - 8 | 899 | 76 | |
CA450619.1 | - | 3 - 4 | 76 |