Assay Design
Gene Transcripts
- Gene Symbol
- TDH
- Entrez Gene ID
-
157739
- Gene Name
- L-threonine dehydrogenase (pseudogene)
- Gene Aliases
-
SDR14E1P
- Chromosome Location
-
Chr.8: 11339637 - 11368452 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX091784.1 |
- |
3 - 4 |
266 |
|
Chr. 8 - 11346178 - 11361742 [+] on Build GRCh38 |
124 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 35 member G5 |
SLC35G5 |
AMAC, AMAC1L2 |
-26.0kb
|
Sense |
Chr. - 11330986 - 11332186 on Build GRCh38 |
Downstream |
FAM167A antisense RNA 1 |
FAM167A-AS1 |
C8orf12 |
9.1kb
|
Sense |
Chr. - 11368402 - 11438657 on Build GRCh38 |
View Full Product Details