Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 22 - 23 | 2882 | 68 |
XM_011513232.2 | XP_011511534.1 | 22 - 23 | 2629 | 68 | |
XM_017007318.1 | XP_016862807.1 | 22 - 23 | 2518 | 68 | |
XM_017007319.1 | XP_016862808.1 | 22 - 23 | 2629 | 68 | |
GenBank mRNA | AK096510.1 | - | 13 - 14 | 1478 | 68 |
AY823162.1 | - | 22 - 23 | 2559 | 68 |