Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 27 - 28 | 3536 | 133 |
XM_017007318.1 | XP_016862807.1 | 27 - 28 | 3172 | 133 | |
GenBank mRNA | AY358110.1 | - | 4 - 5 | 505 | 133 |
AY823162.1 | - | 27 - 28 | 3213 | 133 |