Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 29 - 30 | 3701 | 136 |
XM_017007318.1 | XP_016862807.1 | 29 - 30 | 3337 | 136 | |
GenBank mRNA | AK096510.1 | - | 20 - 21 | 2269 | 136 |
AY358110.1 | - | 6 - 7 | 670 | 136 | |
AY823162.1 | - | 29 - 30 | 3378 | 136 |