Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 10 - 11 | 1432 | 90 |
XM_011513232.2 | XP_011511534.1 | 10 - 11 | 1179 | 90 | |
XM_017007318.1 | XP_016862807.1 | 10 - 11 | 1068 | 90 | |
XM_017007319.1 | XP_016862808.1 | 10 - 11 | 1179 | 90 | |
GenBank mRNA | AK095277.1 | - | 10 - 11 | 1217 | 90 |
AL512736.1 | - | 2 - 3 | 251 | 90 | |
AY823162.1 | - | 10 - 11 | 1109 | 90 | |
BC101496.1 | - | 10 - 11 | 1203 | 90 |