Assay Design



Gene Transcripts

Gene Symbol
TNC
Entrez Gene ID
3371
Gene Name
tenascin C
Gene Aliases
150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
Chromosome Location
Chr.9: 115019575 - 115118257 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002160.3 NP_002151.2 4 - 5 2540 97
XM_005251972.3 XP_005252029.1 4 - 5 2490 97
XM_005251973.3 XP_005252030.1 4 - 5 2490 97
XM_005251974.3 XP_005252031.1 4 - 5 2490 97
XM_005251975.3 XP_005252032.1 4 - 5 2490 97
XM_006717096.3 XP_006717159.1 4 - 5 2490 97
XM_006717097.3 XP_006717160.1 4 - 5 2490 97
XM_006717098.3 XP_006717161.1 4 - 5 2490 97
XM_006717101.3 XP_006717164.1 4 - 5 2490 97
XM_011518625.2 XP_011516927.1 4 - 5 2490 97
XM_011518626.2 XP_011516928.1 4 - 5 2490 97
XM_011518628.2 XP_011516930.1 4 - 5 2490 97
XM_011518629.2 XP_011516931.1 4 - 5 2490 97
XM_017014678.1 XP_016870167.1 4 - 5 2490 97
XM_017014679.1 XP_016870168.1 4 - 5 2490 97
XM_017014680.1 XP_016870169.1 4 - 5 2490 97
XM_017014681.1 XP_016870170.1 4 - 5 2490 97
GenBank mRNA AB210038.1 - 4 - 5 2416 97
AK304015.1 - 4 - 5 2436 97
BC151843.1 - 4 - 5 2408 97
GU473235.1 - 3 - 4 2128 97
M24630.1 - 2 - 3 838 97
M55618.1 - 3 - 4 2263 97
X56160.1 - 3 - 4 2179 97
X78565.1 - 4 - 5 2441 97

View Full Product Details