Assay Design



Gene Transcripts

Gene Symbol
AZI2
Entrez Gene ID
64343
Gene Name
5-azacytidine induced 2
Gene Aliases
AZ2, NAP1, TILP
Chromosome Location
Chr.3: 28322353 - 28349129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005265392.2 XP_005265449.1 2 - 3 615 134

View Full Product Details