Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297418.1 | NP_001284347.1 | 10 - 11 | 1188 | 73 |
NM_005968.4 | NP_005959.2 | 12 - 13 | 1195 | 73 | |
NM_031203.3 | NP_112480.2 | 13 - 14 | 1078 | 73 | |
XM_005272478.2 | XP_005272535.1 | 11 - 12 | 1494 | 73 | |
XM_005272480.2 | XP_005272537.1 | 12 - 13 | 1377 | 73 | |
XM_005272481.1 | XP_005272538.1 | 11 - 12 | 1141 | 73 | |
XM_005272483.2 | XP_005272540.1 | 12 - 13 | 1006 | 73 | |
XM_017026828.1 | XP_016882317.1 | 11 - 12 | 928 | 73 | |
XM_017026831.1 | XP_016882320.1 | 12 - 13 | 993 | 73 | |
XM_017026832.1 | XP_016882321.1 | 11 - 12 | 948 | 73 | |
GenBank mRNA | AB209733.1 | - | 10 - 11 | 1016 | 73 |
AF061832.1 | - | 13 - 14 | 1228 | 73 | |
BC000138.2 | - | 12 - 13 | 1156 | 73 | |
BC008895.2 | - | 3 - 4 | 201 | 73 | |
BC019580.1 | - | 12 - 13 | 1124 | 73 | |
L03532.1 | - | 12 - 13 | 1123 | 73 | |
L32611.1 | - | 13 - 14 | 1007 | 73 | |
X72018.1 | - | 8 - 9 | 761 | 73 |