Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297418.1 | NP_001284347.1 | 13 - 14 | 2103 | 78 |
NM_005968.4 | NP_005959.2 | 15 - 16 | 2110 | 78 | |
NM_031203.3 | NP_112480.2 | 16 - 17 | 1993 | 78 | |
XM_005272478.2 | XP_005272535.1 | 14 - 15 | 2409 | 78 | |
XM_005272479.2 | XP_005272536.1 | 14 - 15 | 2400 | 78 | |
XM_005272480.2 | XP_005272537.1 | 15 - 16 | 2292 | 78 | |
XM_005272481.1 | XP_005272538.1 | 14 - 15 | 2056 | 78 | |
XM_005272483.2 | XP_005272540.1 | 15 - 16 | 1921 | 78 | |
XM_017026823.1 | XP_016882312.1 | 13 - 14 | 2355 | 78 | |
XM_017026824.1 | XP_016882313.1 | 15 - 16 | 2283 | 78 | |
XM_017026825.1 | XP_016882314.1 | 14 - 15 | 2238 | 78 | |
XM_017026826.1 | XP_016882315.1 | 13 - 14 | 1947 | 78 | |
XM_017026827.1 | XP_016882316.1 | 12 - 13 | 1957 | 78 | |
XM_017026828.1 | XP_016882317.1 | 14 - 15 | 1843 | 78 | |
XM_017026829.1 | XP_016882318.1 | 14 - 15 | 1885 | 78 | |
XM_017026830.1 | XP_016882319.1 | 13 - 14 | 1840 | 78 | |
XM_017026831.1 | XP_016882320.1 | 15 - 16 | 1908 | 78 | |
XM_017026832.1 | XP_016882321.1 | 14 - 15 | 1863 | 78 | |
GenBank mRNA | AB209733.1 | - | 13 - 14 | 1931 | 78 |
AF061832.1 | - | 16 - 17 | 2143 | 78 | |
BC000138.2 | - | 15 - 16 | 2071 | 78 | |
BC008895.2 | - | 6 - 7 | 1116 | 78 | |
BC019580.1 | - | 15 - 16 | 2039 | 78 | |
BC064588.1 | - | 2 - 3 | 695 | 78 | |
CN350481.1 | - | 2 - 3 | 253 | 78 | |
CN350489.1 | - | 2 - 3 | 128 | 78 | |
CN350526.1 | - | 2 - 3 | 330 | 78 | |
L03532.1 | - | 15 - 16 | 2038 | 78 | |
L32611.1 | - | 16 - 17 | 1922 | 78 | |
X72018.1 | - | 11 - 12 | 1676 | 78 |