Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145398.2 | NP_001138870.1 | 1 - 2 | 269 | 78 |
GenBank mRNA | AK091916.1 | - | 1 - 2 | 147 | 78 |
DA213449.1 | - | 1 - 2 | 269 | 78 | |
DA243618.1 | - | 1 - 2 | 214 | 78 |