Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004911.4 | NP_004902.1 | 5 - 6 | 1102 | 120 |
XM_006716185.2 | XP_006716248.1 | 5 - 6 | 1102 | 120 | |
GenBank mRNA | AK290971.1 | - | 5 - 6 | 887 | 120 |
AK313299.1 | - | 5 - 6 | 1074 | 120 | |
BC000425.2 | - | 5 - 6 | 864 | 120 | |
BC001928.1 | - | 5 - 6 | 864 | 120 | |
BC006344.2 | - | 5 - 6 | 1052 | 120 | |
BC011754.2 | - | 5 - 6 | 1053 | 120 |