Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008485.1 | NP_001008485.1 | 11 - 12 | 1592 | 75 |
NM_001008486.1 | NP_001008486.1 | 10 - 11 | 1484 | 75 | |
XM_005247559.1 | XP_005247616.1 | 11 - 12 | 1470 | 75 | |
XM_005247562.1 | XP_005247619.1 | 11 - 12 | 1486 | 75 | |
XM_005247563.2 | XP_005247620.1 | 11 - 12 | 1515 | 75 | |
XM_005247565.2 | XP_005247622.1 | 10 - 11 | 1574 | 75 | |
XM_006713681.2 | XP_006713744.1 | 11 - 12 | 1432 | 75 | |
XM_011512945.1 | XP_011511247.1 | 10 - 11 | 1619 | 75 | |
XM_017006705.1 | XP_016862194.1 | 11 - 12 | 1459 | 75 | |
GenBank mRNA | AK000480.1 | - | 10 - 11 | 1315 | 75 |
AK022780.1 | - | 10 - 11 | 1313 | 75 | |
AK128054.1 | - | 6 - 7 | 3626 | 75 | |
AW938624.1 | - | 4 - 5 | 468 | 75 | |
BC009039.2 | - | 11 - 12 | 1425 | 75 | |
BC028241.1 | - | 10 - 11 | 1408 | 75 |