Assay Design



Gene Transcripts

Gene Symbol
SLC41A3
Entrez Gene ID
54946
Gene Name
solute carrier family 41 member 3
Gene Aliases
SLC41A1-L2
Chromosome Location
Chr.3: 126006357 - 126101550 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008485.1 NP_001008485.1 5 - 6 822 65
NM_001008486.1 NP_001008486.1 4 - 5 714 65
NM_001008487.1 NP_001008487.1 4 - 5 741 65
NM_001164475.1 NP_001157947.1 4 - 5 443 65
NM_017836.3 NP_060306.3 5 - 6 822 65
XM_005247559.1 XP_005247616.1 5 - 6 700 65
XM_005247562.1 XP_005247619.1 5 - 6 716 65
XM_005247563.2 XP_005247620.1 5 - 6 745 65
XM_005247564.1 XP_005247621.1 5 - 6 700 65
XM_005247565.2 XP_005247622.1 4 - 5 804 65
XM_006713681.2 XP_006713744.1 5 - 6 662 65
XM_011512945.1 XP_011511247.1 4 - 5 849 65
XM_017006705.1 XP_016862194.1 5 - 6 689 65
XM_017006706.1 XP_016862195.1 5 - 6 716 65
XM_017006707.1 XP_016862196.1 5 - 6 739 65
XM_017006708.1 XP_016862197.1 4 - 5 547 65
XM_017006709.1 XP_016862198.1 4 - 5 602 65
GenBank mRNA AK000480.1 - 4 - 5 545 65
AK021925.1 - 5 - 6 655 65
AK022780.1 - 4 - 5 543 65
AK091671.1 - 6 - 7 626 65
AK093380.1 - 4 - 5 738 65
AK298070.1 - 4 - 5 452 65
AK307107.1 - 5 - 6 744 65
AL560923.3 - 4 - 5 726 65
BC009039.2 - 5 - 6 655 65
BC009444.1 - 5 - 6 640 65
BC028241.1 - 4 - 5 638 65

View Full Product Details