Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000070.2 | NP_000061.1 | 13 - 14 | 2052 | 69 |
NM_024344.1 | NP_077320.1 | 13 - 14 | 2052 | 69 | |
NM_173087.1 | NP_775110.1 | 12 - 13 | 1908 | 69 | |
NM_173088.1 | NP_775111.1 | 2 - 3 | 379 | 69 | |
GenBank mRNA | AF127764.1 | - | 13 - 14 | 1967 | 69 |
AF127765.3 | - | 12 - 13 | 1823 | 69 | |
AK308103.1 | - | 13 - 14 | 2051 | 69 | |
BC003169.1 | - | 2 - 3 | 379 | 69 | |
BC146649.1 | - | 13 - 14 | 1965 | 69 | |
BC146672.1 | - | 13 - 14 | 1965 | 69 | |
BT007322.1 | - | 1 - 2 | 210 | 69 | |
EU791850.1 | - | 13 - 14 | 1485 | 69 | |
EU791851.1 | - | 12 - 13 | 1341 | 69 | |
KU177954.1 | - | 1 - 2 | 210 | 69 | |
KU177955.1 | - | 1 - 2 | 210 | 69 | |
KU177956.1 | - | 1 - 2 | 210 | 69 | |
X85030.1 | - | 13 - 14 | 1746 | 69 |