Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000070.2 | NP_000061.1 | 20 - 21 | 2488 | 66 |
NM_024344.1 | NP_077320.1 | 19 - 20 | 2470 | 66 | |
NM_173087.1 | NP_775110.1 | 17 - 18 | 2212 | 66 | |
NM_173088.1 | NP_775111.1 | 9 - 10 | 815 | 66 | |
NM_173089.1 | NP_775112.1 | 5 - 6 | 445 | 66 | |
NM_173090.1 | NP_775113.1 | 6 - 7 | 463 | 66 | |
GenBank mRNA | AF127764.1 | - | 19 - 20 | 2385 | 66 |
AF127765.3 | - | 17 - 18 | 2127 | 66 | |
AK027249.1 | - | 4 - 5 | 1105 | 66 | |
BC003169.1 | - | 9 - 10 | 815 | 66 | |
BC003521.1 | - | 5 - 6 | 427 | 66 | |
BC004883.2 | - | 5 - 6 | 445 | 66 | |
BC007810.2 | - | 6 - 7 | 411 | 66 | |
BC067126.1 | - | 5 - 6 | 428 | 66 | |
BC100782.1 | - | 5 - 6 | 426 | 66 | |
BC100783.1 | - | 5 - 6 | 426 | 66 | |
BC100784.2 | - | 5 - 6 | 426 | 66 | |
BC107791.1 | - | 6 - 7 | 578 | 66 | |
BC128605.1 | - | 5 - 6 | 426 | 66 | |
BC146649.1 | - | 20 - 21 | 2401 | 66 | |
BC146672.1 | - | 20 - 21 | 2401 | 66 | |
BT007322.1 | - | 8 - 9 | 646 | 66 | |
EU791850.1 | - | 20 - 21 | 1921 | 66 | |
EU791851.1 | - | 19 - 20 | 1777 | 66 | |
KU177954.1 | - | 8 - 9 | 646 | 66 | |
KU177955.1 | - | 8 - 9 | 747 | 66 | |
KU177956.1 | - | 6 - 7 | 514 | 66 | |
X85030.1 | - | 20 - 21 | 2182 | 66 |