Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002976.3 | NP_002967.2 | 16 - 17 | 2827 | 133 |
NR_045628.1 | 17 - 18 | 2921 | 133 | ||
XM_006712680.2 | XP_006712743.1 | 16 - 17 | 2896 | 133 | |
XM_006712681.3 | XP_006712744.1 | 17 - 18 | 3121 | 133 | |
XM_006712682.3 | XP_006712745.1 | 18 - 19 | 3246 | 133 | |
XM_011511615.2 | XP_011509917.1 | 18 - 19 | 3192 | 133 | |
XM_017004667.1 | XP_016860156.1 | 18 - 19 | 3165 | 133 | |
GenBank mRNA | BC141864.1 | - | 17 - 18 | 2923 | 133 |
M91556.1 | - | 16 - 17 | 2836 | 133 |