Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319224.1 | NP_001306153.1 | 8 - 9 | 1421 | 140 |
NM_003686.4 | NP_003677.4 | 7 - 8 | 1258 | 140 | |
NM_006027.4 | NP_006018.4 | 7 - 8 | 1258 | 140 | |
NM_130398.3 | NP_569082.2 | 9 - 10 | 1526 | 140 | |
XM_006711840.2 | XP_006711903.1 | 9 - 10 | 1529 | 140 | |
XM_011544321.1 | XP_011542623.1 | 8 - 9 | 1423 | 140 | |
XM_011544322.1 | XP_011542624.1 | 8 - 9 | 1492 | 140 | |
XM_011544323.2 | XP_011542625.1 | 9 - 10 | 1529 | 140 | |
XM_011544324.2 | XP_011542626.1 | 8 - 9 | 1409 | 140 | |
XM_017002793.1 | XP_016858282.1 | 7 - 8 | 1303 | 140 | |
GenBank mRNA | AF042282.1 | - | 7 - 8 | 1162 | 140 |
AF060479.1 | - | 7 - 8 | 1153 | 140 | |
AF084974.1 | - | 7 - 8 | 1153 | 140 | |
AF091740.1 | - | 7 - 8 | 1155 | 140 | |
AK291291.1 | - | 8 - 9 | 140 | ||
AL080139.1 | - | 3 - 4 | 424 | 140 | |
BC007491.2 | - | 9 - 10 | 1474 | 140 |