Assay Design



Gene Transcripts

Gene Symbol
PHACTR1
Entrez Gene ID
221692
Gene Name
phosphatase and actin regulator 1
Gene Aliases
RPEL, RPEL1, dJ257A7.2
Chromosome Location
Chr.6: 12716635 - 13290484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242648.2 NP_001229577.1 9 - 10 1568 74
NM_001322308.1 NP_001309237.1 8 - 9 1523 74
NM_001322309.1 NP_001309238.1 8 - 9 1414 74
NM_001322310.1 NP_001309239.1 8 - 9 1489 74
NM_001322311.1 NP_001309240.1 6 - 7 1175 74
NM_001322312.1 NP_001309241.1 6 - 7 1538 74
NM_001322313.1 NP_001309242.1 7 - 8 1745 74
NM_001322314.1 NP_001309243.1 7 - 8 1677 74
NM_030948.3 NP_112210.1 9 - 10 1758 74
XM_005248934.2 XP_005248991.1 10 - 11 1807 74
XM_017010452.1 XP_016865941.1 7 - 8 1471 74
XM_017010454.1 XP_016865943.1 10 - 11 1807 74
XM_017010455.1 XP_016865944.1 10 - 11 1775 74
XM_017010456.1 XP_016865945.1 9 - 10 1772 74
XM_017010457.1 XP_016865946.1 9 - 10 1751 74
XM_017010458.1 XP_016865947.1 6 - 7 1264 74
XM_017010459.1 XP_016865948.1 9 - 10 1600 74
XM_017010460.1 XP_016865949.1 9 - 10 1568 74
XM_017010461.1 XP_016865950.1 8 - 9 1539 74
XM_017010462.1 XP_016865951.1 8 - 9 1565 74
XM_017010463.1 XP_016865952.1 7 - 8 1665 74
XM_017010464.1 XP_016865953.1 6 - 7 1264 74
XM_017010465.1 XP_016865954.1 7 - 8 1445 74
XM_017010466.1 XP_016865955.1 7 - 8 1458 74
XM_017010467.1 XP_016865956.1 6 - 7 1445 74
XM_017010469.1 XP_016865958.1 6 - 7 1247 74
GenBank mRNA AB051520.1 - 8 - 9 1365 74
AK290017.1 - 9 - 10 1419 74
AK295263.1 - 10 - 11 1775 74
BC101526.1 - 8 - 9 1332 74
BC101528.1 - 8 - 9 1332 74

View Full Product Details