Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286123.1 | NP_001273052.1 | 2 - 3 | 319 | 78 |
NM_001286125.1 | NP_001273054.1 | 2 - 3 | 319 | 78 | |
NM_005835.3 | NP_005826.1 | 3 - 4 | 769 | 78 | |
XM_005248784.2 | XP_005248841.1 | 3 - 4 | 769 | 78 | |
XM_006714949.3 | XP_006715012.1 | 3 - 4 | 506 | 78 | |
XM_006714950.2 | XP_006715013.1 | 2 - 3 | 396 | 78 | |
XM_006714951.1 | XP_006715014.1 | 3 - 4 | 769 | 78 | |
XM_017010159.1 | XP_016865648.1 | 2 - 3 | 659 | 78 | |
XM_017010160.1 | XP_016865649.1 | 3 - 4 | 764 | 78 | |
GenBank mRNA | BC104822.1 | - | 2 - 3 | 292 | 78 |
BC112033.1 | - | 2 - 3 | 292 | 78 | |
BC143323.1 | - | 2 - 3 | 290 | 78 | |
U90544.1 | - | 3 - 4 | 654 | 78 |