Assay Design



Gene Transcripts

Gene Symbol
SLC17A2
Entrez Gene ID
10246
Gene Name
solute carrier family 17 member 2
Gene Aliases
NPT3
Chromosome Location
Chr.6: 25912754 - 25930727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286123.1 NP_001273052.1 4 - 5 648 66
NM_001286125.1 NP_001273054.1 4 - 5 648 66
NM_005835.3 NP_005826.1 5 - 6 1098 66
XM_005248784.2 XP_005248841.1 5 - 6 1098 66
XM_006714949.3 XP_006715012.1 5 - 6 835 66
XM_006714950.2 XP_006715013.1 4 - 5 725 66
XM_006714951.1 XP_006715014.1 5 - 6 1098 66
XM_017010159.1 XP_016865648.1 4 - 5 988 66
XM_017010160.1 XP_016865649.1 5 - 6 1093 66
GenBank mRNA BC104822.1 - 4 - 5 621 66
BC112033.1 - 4 - 5 621 66
BC143323.1 - 4 - 5 619 66
U90544.1 - 5 - 6 983 66

View Full Product Details