Assay Design



Gene Transcripts

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
Chromosome Location
Chr.17: 44170706 - 44179083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080863.4 NP_543139.4 4 - 5 1255 86
GenBank mRNA AK054727.1 - 4 - 5 1255 86
AK314595.1 - 4 - 5 1247 86
AY557346.1 - 4 - 5 881 86
BC075088.2 - 4 - 5 1204 86
BC118584.1 - 4 - 5 947 86
BC118585.1 - 4 - 5 947 86

View Full Product Details