Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000135.2 | NP_000126.2 | 29 - 30 | 2893 | 80 |
NM_001286167.1 | NP_001273096.1 | 29 - 30 | 2893 | 80 | |
XM_005256294.4 | XP_005256351.1 | 29 - 30 | 2894 | 80 | |
XM_011522946.2 | XP_011521248.1 | 29 - 30 | 2901 | 80 | |
XM_011522947.2 | XP_011521249.1 | 23 - 24 | 2812 | 80 | |
GenBank mRNA | AK299282.1 | - | 29 - 30 | 2882 | 80 |
X99226.1 | - | 29 - 30 | 2882 | 80 |