Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130398.3 | NP_569082.2 | 3 - 4 | 564 | 77 |
XM_006711840.2 | XP_006711903.1 | 3 - 4 | 567 | 77 | |
XM_011544323.2 | XP_011542625.1 | 3 - 4 | 567 | 77 | |
XM_011544324.2 | XP_011542626.1 | 3 - 4 | 567 | 77 | |
GenBank mRNA | BC007491.2 | - | 3 - 4 | 512 | 77 |