Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003592.2 | NP_003583.2 | 11 - 12 | 1650 | 142 |
XM_005250060.4 | XP_005250117.1 | 11 - 12 | 1686 | 142 | |
XM_011516629.2 | XP_011514931.1 | 11 - 12 | 1642 | 142 | |
XM_011516630.2 | XP_011514932.1 | 11 - 12 | 1543 | 142 | |
XM_011516631.2 | XP_011514933.1 | 11 - 12 | 1671 | 142 | |
XM_011516632.2 | XP_011514934.1 | 11 - 12 | 1569 | 142 | |
XM_017012723.1 | XP_016868212.1 | 10 - 11 | 1406 | 142 | |
GenBank mRNA | AF062536.1 | - | 10 - 11 | 1424 | 142 |
AK096163.1 | - | 12 - 13 | 1538 | 142 | |
AK096505.1 | - | 4 - 5 | 140 | ||
AK296909.1 | - | 10 - 11 | 1375 | 142 | |
AK315023.1 | - | 11 - 12 | 1826 | 142 | |
BC125119.1 | - | 11 - 12 | 1563 | 142 | |
BC125120.1 | - | 11 - 12 | 1563 | 142 | |
BX537409.1 | - | 11 - 12 | 1580 | 142 | |
U58087.1 | - | 11 - 12 | 1352 | 142 |