Assay Design



Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
-
Chromosome Location
Chr.7: 148698178 - 148801110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003592.2 NP_003583.2 3 - 4 660 141
XM_005250060.4 XP_005250117.1 3 - 4 696 141
XM_011516629.2 XP_011514931.1 3 - 4 652 141
XM_011516630.2 XP_011514932.1 3 - 4 553 141
XM_011516631.2 XP_011514933.1 3 - 4 681 141
XM_011516632.2 XP_011514934.1 3 - 4 579 141
XM_017012723.1 XP_016868212.1 3 - 4 580 141
GenBank mRNA AF062536.1 - 2 - 3 434 141
AK096163.1 - 4 - 5 548 141
AK315023.1 - 3 - 4 836 141
BC125119.1 - 3 - 4 573 141
BC125120.1 - 3 - 4 573 141
BX537409.1 - 3 - 4 590 141

View Full Product Details