Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015219.4 | NP_056034.2 | 6 - 7 | 899 | 67 |
XM_006721789.3 | XP_006721852.1 | 6 - 7 | 898 | 67 | |
GenBank mRNA | AK023832.1 | - | 6 - 7 | 807 | 67 |
BX647763.1 | - | 5 - 6 | 660 | 67 |